
Tomatine

Synonym(s):
(3β,5α,22β,25S)-Spirosolan-3-yl O-β-D-glucopyranosyl-(1→2)-O-[β-D-xylopyranosyl-(1→3)]-O-β-D-glucopyranosyl-(1→4)-β-D-galactopyranoside, Lycopersicin, NSC 234440, NSC 9223, Spirosolane, α-Tomatine, alpha-tomatine, Lycopersicin
Empirical Formula (Hill Notation):
C50H83NO21
CAS Number:
Molecular Weight:
1034.19
Products
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing