
Titanium diisopropoxide bis(acetylacetonate)

Synonym(s):
Diisopropoxytitanium bis(acetylacetonate) solution, Ti(acac)2OiPr2
Linear Formula:
[(CH3)2CHO]2Ti(C5H7O2)2
CAS Number:
Molecular Weight:
364.26
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing