Skip to Content
Merck
Tetraethylammonium bicarbonate ≥95.0% (T)

Tetraethylammonium bicarbonate

Tetraethylammonium bicarbonate ≥95.0% (T)
Synonym(s):
Tetraethylammonium hydrogen carbonate
Linear Formula:
(CH3CH2)4N(HCO3)
CAS Number:
Molecular Weight:
191.27

Products

Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing