
Somatostatin

Synonym(s):
Growth hormone release inhibiting factor, SRIF, Somatostatin-14, Somatotropin release inhibiting factor
Empirical Formula (Hill Notation):
C76H104N18O19S2
CAS Number:
Molecular Weight:
1637.88
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3