Skip to Content
Merck
Sodium citrate monobasic BioXtra, anhydrous, ≥99.5% (T)

Sodium citrate monobasic

Sodium citrate monobasic BioXtra, anhydrous, ≥99.5% (T)
Synonym(s):
Citric acid monosodium salt, Sodium dihydrogen citrate
Linear Formula:
HOC(COONa)(CH2COOH)2
CAS Number:
Molecular Weight:
214.11
EC Number:

Products

Product Category
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2