Skip to Content
Merck
(S)-(+)-1-Cyclohexylethylamine ChiPros®, produced by BASF, 99%

(S)-(+)-1-Cyclohexylethylamine

(S)-(+)-1-Cyclohexylethylamine ChiPros®, produced by BASF, 99%
Synonym(s):
(S)-(+)-α-Methylcyclohexanemethylamine
Linear Formula:
C6H11CH(CH3)NH2
CAS Number:
Molecular Weight:
127.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing