
Ruthenium(III) acetylacetonate

Synonym(s):
2,4-Pentanedione ruthenium(III) derivative, Ru(acac)3, Ruthenium(III) acetylacetonate
Linear Formula:
(CH3C(O)=CHCOCH3)3Ru
CAS Number:
Molecular Weight:
398.39
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing