
Resorufin β-D-glucopyranoside

Synonym(s):
3-Phenoxazone 7-(β-D-glucopyranoside)
Empirical Formula (Hill Notation):
C18H17NO8
CAS Number:
Molecular Weight:
375.33
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing