
Ramipril Related Compound D

Synonym(s):
Ethyl (2S)2-[(3S,5aS,8aS, 9aS)-3-methyl-1,4-dioxodecahydro-1H-cyclopenta[e]pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate, Ramipril diketopiperazine
Empirical Formula (Hill Notation):
C23H30N2O4
CAS Number:
Molecular Weight:
398.50
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing