
Prochloraz Metabolite BTS44596

Synonym(s):
N′-Formyl-N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]urea
Empirical Formula (Hill Notation):
C13H15Cl3N2O3
CAS Number:
Molecular Weight:
353.63
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing