
Praseodymium

Synonym(s):
Praseodymium
Empirical Formula (Hill Notation):
Pr
CAS Number:
Molecular Weight:
140.91
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Length (mm)
Element
Showing 1-20 of 21
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2