
Polyethyleneglycol Bisphenol A Epichlorohydrin Copolymer

Synonym(s):
Polyethylene glycol compound
Linear Formula:
[C15H16O2·C3H5ClO·(C2H4O)n·H2O]x
CAS Number:
Products
Brand
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing