
PAMAM-succinamic acid dendrimer, 1,4-diaminobutane core, generation 4 solution

Synonym(s):
PAMAM-G4-COOH
Linear Formula:
[NH2(CH2)4NH2]:(G=4);dendri PAMAM(NHCOCH2CH2COOH)64
Molecular Weight:
20646.87
Products
Product Category
Brand
Formula Weight
Physical Form
Generation
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing