
Octaethylene glycol monodecyl ether

Synonym(s):
C10E8, Decyl octaethylene glycol ether, Decyloctaglycol, Polyoxyethylene (8) decyl ether
Linear Formula:
CH3(CH2)9(OCH2CH2)8OH
CAS Number:
Molecular Weight:
510.70
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing