Skip to Content
Merck
Noratropine phyproof® Reference Substance

Noratropine

Noratropine phyproof® Reference Substance
Synonym(s):
N-Demethylatropine
Empirical Formula (Hill Notation):
C16H21NO3
CAS Number:
Molecular Weight:
275.34

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing