Skip to Content
Merck
N,N-Dimethyldodecylamine N-oxide solution ~30% in H2O

N,N-Dimethyldodecylamine N-oxide solution

N,N-Dimethyldodecylamine N-oxide solution ~30% in H2O
Synonym(s):
DDAO, LDAO, Lauryldimethylamine N-oxide
Empirical Formula (Hill Notation):
C14H31NO
CAS Number:
Molecular Weight:
229.40

Products

Product Category
Formula Weight
Physical Form
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing