
Nε-Trifluoroacetyl-L-lysine

Synonym(s):
ε-TFA-lysine
Empirical Formula (Hill Notation):
C8H13F3N2O3
CAS Number:
Molecular Weight:
242.20
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing