![N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala](/deepweb/assets/sigmaaldrich/product/structures/805/876/96b5fb57-71c8-4c6b-b5d2-fafe7374cd85/640/96b5fb57-71c8-4c6b-b5d2-fafe7374cd85.png)
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala
![N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala](/deepweb/assets/sigmaaldrich/product/structures/805/876/96b5fb57-71c8-4c6b-b5d2-fafe7374cd85/640/96b5fb57-71c8-4c6b-b5d2-fafe7374cd85.png)
Synonym(s):
FALGPA, N-[3-(2-Furyl)acryloyl]-L-leucyl-glycyl-L-prolyl-L-alanine
Empirical Formula (Hill Notation):
C23H32N4O7
CAS Number:
Molecular Weight:
476.52
Products
Product Category
Brand
Formula Weight
Gene Alias
Showing 1-1 of 1
Compare
Product Number
Description
Pricing