
Morphine sulfate salt pentahydrate

Synonym(s):
Morphine hemi[sulfate pentahydrate]
Empirical Formula (Hill Notation):
C34H40N2O10S · 5H2O
CAS Number:
Molecular Weight:
758.83
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing