
modified Tryptic Soy Broth- Dehydrated Culture Media

Products
Product Category
Brand
Color
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing