
Methyl 3-aminocrotonate

Synonym(s):
Methyl 3-amino-2-butenoate
Linear Formula:
CH3C(NH2)=CHCOOCH3
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing