
Metamizole Impurity A

Synonym(s):
4-Formylaminoantipyrine, N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide
Empirical Formula (Hill Notation):
C12H13N3O2
CAS Number:
Molecular Weight:
231.25
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2