Skip to Content
BS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium oxide
Synonym(s):
Magnesium oxide
Linear Formula:
MgO
CAS Number:
1309-48-4
Molecular Weight:
40.30
EC Number:
215-171-9
Products
Product Category
salts (8)
oxides (2)
Brand
SAFC (2)
Sigma-Aldrich (11)
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reagent Type
Core
Reference Material Type
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
243388
Magnesium oxide,
ACS reagent, 97%
Expand
View Pricing
13138
Magnesium oxide,
puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Expand
View Pricing
342793
Magnesium oxide,
≥99% trace metals basis, -325 mesh
Expand
View Pricing
549649
Magnesium oxide,
nanopowder, ≤50 nm particle size (BET)
Expand
View Pricing
529699
Magnesium oxide,
≥99.99% trace metals basis
Expand
View Pricing
307742
Magnesium oxide,
light, 95%
Expand
View Pricing
1.05862
Magnesium oxide,
light extra pure, Ph. Eur., BP, USP, E 530
Expand
View Pricing
1.05865
Magnesium oxide,
for analysis EMSURE
®
Expand
View Pricing
220361
Magnesium oxide,
-10-+50 mesh, 98%
Expand
View Pricing
1.05867
Magnesium oxide,
heavy, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, ChP, USP
Expand
View Pricing
203718
Magnesium oxide,
99.99% trace metals basis
Expand
View Pricing
1.05866
Magnesium oxide,
for analysis (max.0.001% SO₄) EMSURE
®
ACS
Expand
View Pricing
634646
Magnesium oxide,
(single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Expand
View Pricing
1374281
Magnesium oxide,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
63089
Magnesium oxide,
BioUltra, ≥97.0% (calcined substance, KT)
Expand
View Pricing
63107
Magnesium oxide,
tested according to Ph. Eur., heavy
Expand
View Pricing