Skip to Content
Merck
L-Threoninol 97%

L-Threoninol

L-Threoninol 97%
Synonym(s):
(2R,3R)-2-Amino-1,3-butanediol
Linear Formula:
CH3CH(OH)CH(NH2)CH2OH
CAS Number:
Molecular Weight:
105.14

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing