
Lisinopril S,S,S-diketopiperazine

Synonym(s):
(S)-2-{(3S,8aS)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(1H)-yl}-4-phenylbutanoic acid
Empirical Formula (Hill Notation):
C21H29N3O4
CAS Number:
Molecular Weight:
387.47
Products
Brand
Formula Weight
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing