
Licarbazepine

Synonym(s):
10,11-Dihydro-10-hydroxycarbamazepine, 10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, Licarbazepine
Empirical Formula (Hill Notation):
C15H14N2O2
CAS Number:
Molecular Weight:
254.28
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing