
Levomedetomidine

Synonym(s):
(R)-4-[1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole monohydrochloride
Empirical Formula (Hill Notation):
C13H16N2 · HCl
CAS Number:
Molecular Weight:
236.74
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2