Skip to Content
Merck
Lactulose British Pharmacopoeia (BP) Reference Standard

Lactulose

Lactulose British Pharmacopoeia (BP) Reference Standard
Synonym(s):
4-O-β-D-Galactopyranosyl-D-fructofuranose, 4-O-β-D-Galactopyranosyl-D-fructose, Lactulose
Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique