
(−)-Ipc2B(allyl)borane solution

Synonym(s):
(−)-B-Allyldiisopinocampheylborane solution
Linear Formula:
(C10H17)2BCH2CH=CH2
CAS Number:
Molecular Weight:
326.37
Products
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing