
Indomethacin

Synonym(s):
1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-3-indoleacetic acid, Indomethacin
Empirical Formula (Hill Notation):
C19H16ClNO4
CAS Number:
Molecular Weight:
357.79
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing