
Indole-3-butyric acid potassium salt

Synonym(s):
4-(3-Indolyl)butanoic acid, IBA
Empirical Formula (Hill Notation):
C12H12KNO2
CAS Number:
Molecular Weight:
241.33
Products
Brand
Formula Weight
Product Line
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing