Skip to Content
BS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hydrogen peroxide 30%
Synonym(s):
Hydrogen peroxide solution
Empirical Formula (Hill Notation):
H
2
O
2
CAS Number:
7722-84-1
Molecular Weight:
34.01
Products
Brand
Supelco (2)
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.07209
Hydrogen peroxide 30%,
(Perhydrol
™
) for analysis EMSURE ISO
Expand
View Pricing
1.07210
Hydrogen peroxide 30%,
(Perhydrol
™
) (stabilized for higher storage temp.) for analysis EMSURE ISO
Expand
View Pricing
1.93007
Hydrogen peroxide 30%,
for analysis EMPARTA
®
ACS
Expand
View Pricing