Skip to Content
Merck
HS157 Umicore, 97%

HS157

HS157 Umicore, 97%
Synonym(s):
Bis(1,5-cyclooctadiene)diiridium(I) dichloride, 1,5-Cyclooctadiene-iridium(I) chloride dimer, Chloro(1,5-cyclooctadiene)iridium(I) dimer, Di-μ-chlorobis[(1,2,5,6-η)-1,5-cyclooctadiene]diiridium, Iridium(I) chloride 1,5-cyclooctadiene complex dimer, [Ir(1,5-cod)Cl]2, [Ir(cod)Cl]2
Empirical Formula (Hill Notation):
C16H24Cl2Ir2
CAS Number:
Molecular Weight:
671.70
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing