Skip to Content
Merck
holo-Transferrin bovine

holo-Transferrin bovine

holo-Transferrin bovine
Synonym(s):
Siderophilin, Siderophilin, iron-saturated
CAS Number:
EC Number:

Products

Product Category
Brand
Biological Source
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-2 of 2