Skip to Content
BS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexamethylenediamine
Synonym(s):
1,6-Diaminohexane, 1,6-Hexanediamine, HMDA
Linear Formula:
NH
2
(CH
2
)
6
NH
2
CAS Number:
124-09-4
Molecular Weight:
116.20
EC Number:
204-679-6
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technical (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
H11696
Hexamethylenediamine,
98%
Expand
View Pricing
422002
Hexamethylenediamine,
technical grade, 70%
Expand
View Pricing
52003
Hexamethylenediamine,
analytical standard
Expand
View Pricing
13-0730
Hexamethylenediamine,
SAJ first grade, ≥98.0%
Expand
View Pricing