
Glycolic acid nitrile solution

Synonym(s):
Formaldehyde cyanohydrin, Hydroxyacetonitrile
Linear Formula:
HOCH2CN
CAS Number:
Molecular Weight:
57.05
Products
Product Category
Brand
Formula Weight
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing