Flumequine
Empirical Formula (Hill Notation):
C14H12FNO3
CAS Number:
Molecular Weight:
261.25
Products
Formula Weight
Manufacturer
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing