
Fluconazole Impurity G

Synonym(s):
1-[[(2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl]methyl]-1H-1,2,4-triazole
Empirical Formula (Hill Notation):
C11H9F2N3O
CAS Number:
Molecular Weight:
237.21
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing