
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride

Empirical Formula (Hill Notation):
C8H6O4
CAS Number:
Molecular Weight:
166.13
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing