
Etridiazole

Synonym(s):
5-Ethoxy-3-trichloromethyl-1,2,4-thiadiazole, Ethazol
Empirical Formula (Hill Notation):
C5H5Cl3N2OS
CAS Number:
Molecular Weight:
247.53
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2