
Eriodictyol

Synonym(s):
3′,4′,5,7-Tetrahydroxyflavanone, 3·,4·,5,7-Tetrahydroxyflavanone
Empirical Formula (Hill Notation):
C15H12O6
CAS Number:
Molecular Weight:
288.25
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing