
Epoxy embedding medium, hardener MNA

Synonym(s):
Methyl-5-norbornene-2,3-dicarboxylic anhydride, Methyl nadic anhydride
Empirical Formula (Hill Notation):
C10H10O3
Products
Biological Source
Formula Weight
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing