
(+)-N-Boc-L-α-phenylglycinol

Synonym(s):
(S)-(+)-2-(Boc-amino)-2-phenylethanol
Linear Formula:
(CH3)3CO2CNHCH(C6H5)CH2OH
CAS Number:
Molecular Weight:
237.29
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing