
DMT-dC(tac) Phosphoramidite

Synonym(s):
DMT-dC(tac) amidite, N-[2-[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C51H62N5O9P
CAS Number:
Molecular Weight:
920.04
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Technique