Skip to Content
Merck
Dichlorophenylborane 97%

Dichlorophenylborane

Dichlorophenylborane 97%
Synonym(s):
NSC 93889, Phenylboron dichloride, Phenyldichloroborane
Empirical Formula (Hill Notation):
C6H5BCl2
CAS Number:
Molecular Weight:
158.82

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing