
Diallyldimethylammonium chloride solution

Synonym(s):
Dimethyldiallylammonium chloride
Linear Formula:
[(CH3)2N+(CH2CH=CH2)2]Cl-
CAS Number:
Molecular Weight:
161.67
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing