
Cyproterone acetate for peak identification

Synonym(s):
Cyproterone acetate, 6-Chloro-1β,2β-dihydro-17-hydroxy-3′H-cyclopropa(1,2)-pregna-1,4,6-triene-3,20-dione acetate
Empirical Formula (Hill Notation):
C24H29ClO4
CAS Number:
Molecular Weight:
416.94
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing