
Cyazofamid

Synonym(s):
4-Chloro-1-(dimethylaminosulfonyl)-5-(p-tolyl)imidazole-2-carbonitrile
Empirical Formula (Hill Notation):
C13H13ClN4O2S
CAS Number:
Molecular Weight:
324.79
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2