Skip to Content
BS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chromium(VI) oxide
Synonym(s):
Chromic anhydride, Chromium(VI) oxide
Empirical Formula (Hill Notation):
CrO
3
CAS Number:
1333-82-0
Molecular Weight:
99.99
EC Number:
215-607-8
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (5)
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Special Grade
ACS reagent (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
236470
Chromium(VI) oxide,
ACS reagent, ≥98.0%
Expand
View Pricing
232653
Chromium(VI) oxide,
ReagentPlus
®
, 99.9% trace metals basis
Expand
View Pricing
675644
Chromium(VI) oxide,
99.99% trace metals basis
Expand
View Pricing
8.22266
Chromium(VI) oxide,
for synthesis
Expand
View Pricing
1.00229
Chromium(VI) oxide,
for analysis EMSURE
®
Expand
View Pricing
1.94963
Chromium(VI) oxide,
EMPLURA
®
Expand
View Pricing
05-4600
Chromium(VI) oxide,
SAJ special grade, ≥98.0%
Expand
View Pricing