
Chromium isotopic standard

Synonym(s):
Chromium(III) nitrate nonahydrate
Linear Formula:
Cr(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
400.15
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Isotope
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing